ENA413174413 Enamine

1,3-dibromo-2-methylpropane

  • MDL number MFCD18909351
  • Cas Number : 28148-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85196
Availability details for
Quantity
  • Availability
  • Add to Cart