ENA413174414 Enamine

2,5-dibromohexanedioic acid

  • MDL number MFCD00046516
  • Cas Number : 3479-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85360

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-85360
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart