ENA413174459 Enamine

3-methyl-1,2-oxazole

  • MDL number MFCD00020814
  • Cas Number : 30842-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-88524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88524
Availability details for
Quantity
  • Availability
  • Add to Cart