ENA413174470 Enamine

1-(oxan-4-yl)propan-2-amine

  • MDL number MFCD16745941
  • Cas Number : 854697-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90406
Availability details for
Quantity
  • Availability
  • Add to Cart