ENA413174471 Enamine

methyl 1-nitronaphthalene-2-carboxylate

  • MDL number MFCD06203942
  • Cas Number : 78508-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90442
Availability details for
Quantity
  • Availability
  • Add to Cart