ENA413174498 Enamine

2-(2-fluorophenyl)azetidine

  • MDL number MFCD18702313
  • Cas Number : 1270489-49-0
  • Molecular Weight : 151.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-fluorophenyl)azetidine
Inchi Code 1S/C9H10FN/c10-8-4-2-1-3-7(8)9-5-6-11-9/h1-4,9,11H,5-6H2
Inchi Key YZPUUVPTLJZGDR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-90866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90866
Availability details for
Quantity
  • Availability
  • Add to Cart