ENA413174514 Enamine

4-(3-aminophenoxy)-N-methylpyridine-2-carboxamide

  • MDL number MFCD08447094
  • Cas Number : 284462-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91326
Availability details for
Quantity
  • Availability
  • Add to Cart