ENA413174515 Enamine

2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

  • MDL number MFCD03426404
  • Cas Number : 1127-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...71
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91346

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-91346
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart