ENA413174525 Enamine

3-(2-methyl-1H-1,3-benzodiazol-1-yl)propanoic acid

  • MDL number MFCD01017150
  • Cas Number : 30163-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91803
Availability details for
Quantity
  • Availability
  • Add to Cart