ENA413174553 Enamine

4-(bromomethyl)-2-chloroquinoline

  • MDL number MFCD12910754
  • Cas Number : 137354-54-2
  • Molecular Weight : 256.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-93770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93770
Availability details for
Quantity
  • Availability
  • Add to Cart