ENA413174620 Enamine

ethyl 3-(4-bromo-1H-pyrazol-1-yl)-2-methylpropanoate

  • MDL number MFCD16087643
  • Cas Number : 1249644-53-8
  • Molecular Weight : 261.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96783
Availability details for
Quantity
  • Availability
  • Add to Cart