ENA413174621 Enamine

[(2-chloropyridin-4-yl)methyl]dimethylamine

  • MDL number MFCD13249513
  • Cas Number : 933689-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96897
Availability details for
Quantity
  • Availability
  • Add to Cart