ENA413174625 Enamine

1-(dimethylamino)cyclopentane-1-carbonitrile

  • MDL number MFCD09880530
  • Cas Number : 22912-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97016
Availability details for
Quantity
  • Availability
  • Add to Cart