ENA423199817 Enamine

2-(3a,4,5,6,7,7a-hexahydro-1H-1,3-benzodiazol-2-yl)acetonitrile

  • MDL number MFCD27980623
  • Cas Number : 1803570-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125442

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125442
Availability details for
Quantity
  • Availability
  • Add to Cart