ENA423199825 Enamine

5-(difluoromethyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD27959349
  • Cas Number : 1597679-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138931
Availability details for
Quantity
  • Availability
  • Add to Cart