ENA423199827 Enamine

4-(4-chloro-3-methoxyphenyl)-4-oxobutanoic acid

  • MDL number MFCD27980632
  • Cas Number : 1803599-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141310
Availability details for
Quantity
  • Availability
  • Add to Cart