ENA423199829 Enamine

benzyl[(2S)-3-methoxybutan-2-yl]amine

  • MDL number MFCD27959352
  • Cas Number : 1807882-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142056
Availability details for
Quantity
  • Availability
  • Add to Cart