ENA423199831 Enamine

3,4-diethyl 1-methyl 6-ethoxy-1,4,5,6-tetrahydropyridazine-1,3,4-tricarboxylate

  • MDL number MFCD27980637
  • Cas Number : 1803580-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152949
Availability details for
Quantity
  • Availability
  • Add to Cart