ENA423199836 Enamine

tert-butyl 2-[3-(dimethylamino)-1,2,4-oxadiazol-5-yl]pyrrolidine-1-carboxylate

  • MDL number MFCD27980641
  • Cas Number : 1803610-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-153585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153585
Availability details for
Quantity
  • Availability
  • Add to Cart