ENA423199837 Enamine

tert-butyl N-{2-[3-(dimethylamino)-1,2,4-oxadiazol-5-yl]ethyl}carbamate

  • MDL number MFCD27980642
  • Cas Number : 1803599-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-153586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153586
Availability details for
Quantity
  • Availability
  • Add to Cart