ENA423199841 Enamine

methyl 2-phenyl-3-sulfopropanoate

  • MDL number MFCD27980645
  • Cas Number : 1803560-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-154448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154448
Availability details for
Quantity
  • Availability
  • Add to Cart