ENA423199842 Enamine

5-({[(tert-butoxy)carbonyl]amino}methyl)-4,5-dihydro-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD27959357
  • Cas Number : 1785259-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154513
Availability details for
Quantity
  • Availability
  • Add to Cart