ENA423199844 Enamine

6-hydroxypyrimidine-4-carbonitrile

  • MDL number MFCD27980650
  • Cas Number : 1192064-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155352
Availability details for
Quantity
  • Availability
  • Add to Cart