ENA423199845 Enamine

4-bromo-2-methyl-1-benzofuran-7-carboxylic acid

  • MDL number MFCD27959362
  • Cas Number : 1797928-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155385
Availability details for
Quantity
  • Availability
  • Add to Cart