ENA423199846 Enamine

4-chloro-1-[(1-methoxypropan-2-yl)oxy]-2-nitrobenzene

  • MDL number MFCD27980651
  • Cas Number : 1803590-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155485

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155485
Availability details for
Quantity
  • Availability
  • Add to Cart