ENA423199847 Enamine

3-[methyl(piperidin-4-yl)amino]propanoic acid dihydrochloride

  • MDL number MFCD27980652
  • Cas Number : 1797017-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155508

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155508
Availability details for
Quantity
  • Availability
  • Add to Cart