ENA423199850 Enamine

(2R)-2-amino-N,3-diphenylpropanamide hydrochloride

  • MDL number MFCD27959366
  • Cas Number : 1416505-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155767
Availability details for
Quantity
  • Availability
  • Add to Cart