ENA423199853 Enamine

N'-(5-aminopyridin-2-yl)-2-methylpropanehydrazide

  • MDL number MFCD27959370
  • Cas Number : 1803590-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156155

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156155
Availability details for
Quantity
  • Availability
  • Add to Cart