ENA423199855 Enamine

N-methanesulfonyl-N-(3-nitrophenyl)methanesulfonamide

  • MDL number MFCD27980655
  • Cas Number : 32763-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156220
Availability details for
Quantity
  • Availability
  • Add to Cart