ENA423199858 Enamine

{[amino(pyridin-4-yl)methylidene]amino}urea

  • Cas Number : 1807938-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156485
Availability details for
Quantity
  • Availability
  • Add to Cart