ENA423199867 Enamine

methyl 3-hydroxy-4-[2-(trimethylsilyl)ethynyl]benzoate

  • MDL number MFCD27959379
  • Cas Number : 478169-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157235
Availability details for
Quantity
  • Availability
  • Add to Cart