ENA423199872 Enamine

1-{[(3-methyloxetan-3-yl)methyl]sulfanyl}ethan-1-one

  • MDL number MFCD27980667
  • Cas Number : 1803588-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157484

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157484
Availability details for
Quantity
  • Availability
  • Add to Cart