ENA423199875 Enamine

(1R)-1-(5-chloropyridin-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD27980670
  • Cas Number : 1807920-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157611
Availability details for
Quantity
  • Availability
  • Add to Cart