ENA423199876 Enamine

sodium 5-methyl-2-(4H-1,2,4-triazol-4-yl)benzoate

  • MDL number MFCD27959384
  • Cas Number : 1796952-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157656

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157656
Availability details for
Quantity
  • Availability
  • Add to Cart