ENA423199877 Enamine

methyl 1H,2H,3H,4H,5H-pyrido[4,3-b]indole-8-carboxylate hydrochloride

  • MDL number MFCD27959385
  • Cas Number : 1803588-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157663
Availability details for
Quantity
  • Availability
  • Add to Cart