ENA423199879 Enamine

2-phenylpyrimidine-5-sulfonyl chloride

  • MDL number MFCD27980676
  • Cas Number : 1803607-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157720

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157720
Availability details for
Quantity
  • Availability
  • Add to Cart