ENA423199880 Enamine

[2-(methylamino)pyridin-4-yl]methanol hydrochloride

  • MDL number MFCD27980678
  • Cas Number : 1803593-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157757
Availability details for
Quantity
  • Availability
  • Add to Cart