ENA423199884 Enamine

2-(oxolan-2-yl)ethyl methanesulfonate

  • MDL number MFCD27980682
  • Cas Number : 133243-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157805
Availability details for
Quantity
  • Availability
  • Add to Cart