ENA423199899 Enamine

1-(3-bromopropyl)-4-(propan-2-yl)piperazine dihydrobromide

  • MDL number MFCD27959392
  • Cas Number : 1803593-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-158072
Availability details for
Quantity
  • Availability
  • Add to Cart