ENA423199903 Enamine

methyl 4-(chlorosulfonyl)-3-methoxybenzoate

  • MDL number MFCD27959393
  • Cas Number : 162141-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158091
Availability details for
Quantity
  • Availability
  • Add to Cart