ENA423199904 Enamine

{[1,2,4]triazolo[1,5-a]pyridin-2-yl}hydrazine

  • MDL number MFCD27959394
  • Cas Number : 1693854-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-158459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-158459
Availability details for
Quantity
  • Availability
  • Add to Cart