ENA423199913 Enamine

(1H-imidazol-2-yl)(piperidin-4-yl)methanol

  • MDL number MFCD27980717
  • Cas Number : 1785022-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159710
Availability details for
Quantity
  • Availability
  • Add to Cart