ENA423199917 Enamine

5-(aminomethyl)-4,5-dihydro-1,2-oxazole-3-carboxamide hydrochloride

  • MDL number MFCD27959403
  • Cas Number : 1796992-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159783

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159783
Availability details for
Quantity
  • Availability
  • Add to Cart