ENA423199920 Enamine

2-[(4-aminocyclohexyl)oxy]pyridine-3-carbonitrile hydrochloride

  • MDL number MFCD27959408
  • Cas Number : 1803583-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159883

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159883
Availability details for
Quantity
  • Availability
  • Add to Cart