ENA423199924 Enamine

2-(2-aminoethyl)-6-bromophenol hydrobromide

  • MDL number MFCD27959412
  • Cas Number : 1795187-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-159933
Availability details for
Quantity
  • Availability
  • Add to Cart