ENA423199926 Enamine

1-(2-fluorophenyl)-1,4-diazepane hydrochloride

  • MDL number MFCD27959415
  • Cas Number : 1803611-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159942
Availability details for
Quantity
  • Availability
  • Add to Cart