ENA423199947 Enamine

methyl 6-(2-chloroacetamido)pyridine-2-carboxylate

  • MDL number MFCD27980727
  • Cas Number : 1803601-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160102
Availability details for
Quantity
  • Availability
  • Add to Cart