ENA423199950 Enamine

2-oxabicyclo[4.2.0]octan-7-amine hydrochloride

  • MDL number MFCD27959441
  • Cas Number : 1797303-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160175
Availability details for
Quantity
  • Availability
  • Add to Cart