ENA423199959 Enamine

1-(5-bromo-2-hydroxyphenyl)-2-(2-fluorophenyl)ethan-1-one

  • MDL number MFCD27959450
  • Cas Number : 1300581-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160284
Availability details for
Quantity
  • Availability
  • Add to Cart