ENA423199960 Enamine

1-(5-bromo-2-hydroxyphenyl)-2-phenylethan-1-one

  • MDL number MFCD27959451
  • Cas Number : 54981-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160285
Availability details for
Quantity
  • Availability
  • Add to Cart